siRNA Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271443.1 | NP_001258372.1 | 4 | 226 | ||
| NM_001271444.1 | NP_001258373.1 | 4 | 466 | |||
| NM_144870.5 | NP_659119.2 | 4 | 469 | |||
| GenBank mRNA | AK135069.1 | - | 4 | 227 | ||
| AK149674.1 | - | 4 | 427 | |||
| AK153182.1 | - | 4 | 413 | |||
| AK153900.1 | - | 4 | 203 | |||
| AK164901.1 | - | 4 | 203 | |||
| AK167907.1 | - | 4 | 204 | |||
| BC086766.1 | - | 4 | 180 | 6820786 | ||

Ndufs8
Genomic Map