siRNA Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177798.1 | NP_001171269.1 | 12 | 1661 | ||
| NM_146092.2 | NP_666204.2 | 12 | 1498 | |||
| GenBank mRNA | AK077149.1 | - | 12 | 1662 | ||
| AK188124.1 | - | 1 | 63 | |||
| BC020299.1 | - | 12 | 1471 | |||
| BC028647.1 | - | 12 | 1445 | 1244243 | ||

Taf6l
Genomic Map