siRNA Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177798.1 | NP_001171269.1 | 9 | 1007 | ||
| NM_146092.2 | NP_666204.2 | 9 | 844 | |||
| GenBank mRNA | AK077149.1 | - | 9 | 1008 | ||
| AK144881.1 | - | 9 | 811 | |||
| BC020299.1 | - | 9 | 817 | |||
| BC028647.1 | - | 9 | 791 | 1244243 | ||

Taf6l
Genomic Map