siRNA Details
| Gene Symbol: Ubxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146093.1 | NP_666205.1 | 8 | 692 | ||
| GenBank mRNA | AK150442.1 | - | 7 | 695 | ||
| AK159708.1 | - | 7 | 667 | |||
| AK172428.1 | - | 3 | 1427 | |||
| BC006701.1 | - | 8 | 692 | 3583916 | ||

Ubxn1
Genomic Map