siRNA Details
| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145497.2 | NP_663472.1 | 4 | 239 | ||
| GenBank mRNA | AK050579.1 | - | 4 | 270 | ||
| AK085391.1 | - | 1 | 1553 | |||
| AK169962.1 | - | 4 | 213 | |||
| BC016495.1 | - | 4 | 239 | 4221437 | ||

Nmrk1
Genomic Map