siRNA Details
| Gene Symbol: D030056L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177640.4 | NP_808308.1 | 2 | 533 | ||
| GenBank mRNA | AK083631.1 | - | 2 | 534 | ||
| AK145019.1 | - | 2 | 581 | |||
| AK155389.1 | - | 2 | 574 | |||
| AK213250.1 | - | 1 | 141 | |||
| BC020125.1 | - | 2 | 514 | 3985075 | ||

D030056L22Rik
Genomic Map