siRNA Details
| Gene Symbol: Smc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252684.1 | NP_001239613.1 | 10, 11 | 1538 | ||
| NM_001252685.1 | NP_001239614.1 | 10, 11 | 1367 | |||
| NM_153808.2 | NP_722503.1 | 10, 11 | 1538 | |||
| GenBank mRNA | AJ534941.1 | - | 10, 11 | 1459 | ||
| AK122321.1 | - | 3, 4 | 484 | |||
| AK143818.1 | - | 10, 11 | 1516 | |||
| AK161254.1 | - | 10, 11 | 1368 | |||
| BC007174.1 | - | 1, 2 | 94 | 3493165 | ||
| BC038345.1 | - | 10, 11 | 1518 | 3596405 | ||

Smc5
Genomic Map