siRNA Details
| Gene Symbol: Smc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252684.1 | NP_001239613.1 | 23 | 3128 | ||
| NM_001252685.1 | NP_001239614.1 | 22 | 2915 | |||
| NM_153808.2 | NP_722503.1 | 22 | 3086 | |||
| GenBank mRNA | AJ534941.1 | - | 23 | 3049 | ||
| AK122321.1 | - | 15 | 5279 | |||
| AK161254.1 | - | 22 | 2918 | |||
| BC038345.1 | - | 22 | 3066 | 3596405 | ||

Smc5
Genomic Map