siRNA Details
| Gene Symbol: Cbwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146097.3 | NP_666209.1 | 5, 6 | 578 | ||
| GenBank mRNA | AK076027.1 | - | 5, 6 | 533 | ||
| AK080654.1 | - | 1, 2 | 59 | |||
| AK132291.1 | - | 5, 6 | 564 | |||
| BC018472.1 | - | 5, 6 | 520 | 3661842 | ||
| BC053745.1 | - | 5, 6 | 613 | 6505520 | ||

Cbwd1
Genomic Map