siRNA Details
| Gene Symbol: Dmrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145831.3 | NP_665830.1 | 4 | 1226 | ||
| GenBank mRNA | AF080623.1 | - | 197 | |||
| AF539811.1 | - | 4 | 1226 | |||
| AK044891.1 | - | 4 | 1227 | |||
| AK140337.1 | - | 4 | 1218 | |||
| AK140347.1 | - | 4 | 1221 | |||
| BC027669.1 | - | 1 | 175 | 1248080 | ||

Dmrt2
Genomic Map