siRNA Details
| Gene Symbol: Afap1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177796.1 | NP_001171267.1 | 9, 10 | 1338 | ||
| NM_001177797.1 | NP_001171268.1 | 8, 9 | 1284 | |||
| NM_146102.2 | NP_666214.1 | 10, 11 | 1390 | |||
| GenBank mRNA | AK048649.1 | - | 10, 11 | 1044 | ||
| AK053222.1 | - | 12, 13 | 1640 | |||
| AK087449.1 | - | 9, 10 | 974 | |||
| AK220430.1 | - | 8, 9 | 907 | |||
| BC031515.1 | - | 9, 10 | 940 | 3711746 | ||

Afap1l2
Genomic Map