siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145505.4 | NP_663480.3 | 10 | 1683 | ||
| GenBank mRNA | AK029835.1 | - | 10 | 2013 | ||
| AK046518.1 | - | 10 | 1676 | |||
| AK054539.1 | - | 10 | 1631 | |||
| AK122530.1 | - | 8 | 1162 | |||
| AK147386.1 | - | 8 | 1173 | |||
| BC023048.1 | - | 1 | 155 | |||
| BC062949.1 | - | 10 | 1422 | 6839812 | ||
| BC079562.1 | - | 3 | 274 | |||

Fam160b1
Genomic Map