siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145505.4 | NP_663480.3 | 11 | 1818 | ||
| GenBank mRNA | AK029835.1 | - | 11 | 2148 | ||
| AK046518.1 | - | 11 | 1811 | |||
| AK122530.1 | - | 9 | 1297 | |||
| AK147386.1 | - | 9 | 1308 | |||
| AK172373.1 | - | 11 | 1783 | |||
| BC023048.1 | - | 2 | 290 | |||
| BC062949.1 | - | 11 | 1557 | 6839812 | ||
| BC079562.1 | - | 4 | 409 | |||

Fam160b1
Genomic Map