siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145505.4 | NP_663480.3 | 14 | 2185 | ||
| GenBank mRNA | AK029835.1 | - | 14 | 2515 | ||
| AK046518.1 | - | 14 | 2178 | |||
| AK122530.1 | - | 12 | 1664 | |||
| AK147386.1 | - | 12 | 1675 | |||
| AK172373.1 | - | 14 | 2150 | |||
| BC023048.1 | - | 5 | 657 | |||
| BC062949.1 | - | 14 | 1883 | 6839812 | ||
| BC079562.1 | - | 7 | 776 | |||

Fam160b1
Genomic Map