siRNA Details
| Gene Symbol: Prlhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201615.2 | NP_963909.2 | 1 | 647 | ||
| GenBank mRNA | BC125649.1 | - | 1 | 579 | 40130164 | |
| BC125651.1 | - | 1 | 579 | 40130166 | ||
More Information
Set Membership: |
|

Prlhr
Genomic Map