siRNA Details
| Gene Symbol: 5430435G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145509.2 | NP_663484.1 | 4 | 422 | ||
| GenBank mRNA | AB232641.1 | - | 3 | 266 | ||
| AK030688.1 | - | 4 | 422 | |||
| AK036056.1 | - | 4 | 411 | |||
| AK079999.1 | - | 4 | 390 | |||
| CJ137398.1 | - | 4 | 341 | |||
| JX431303.1 | - | 3 | 266 | |||

5430435G22Rik
Genomic Map