siRNA Details
| Gene Symbol: Rab29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144875.2 | NP_659124.2 | 5, 6 | 772 | ||
| GenBank mRNA | AB232625.1 | - | 4, 5 | 505 | ||
| AK079752.1 | - | 4, 5 | 626 | |||
| BC016133.1 | - | 5, 6 | 757 | 4485029 | ||
| BC029056.1 | - | 4, 5 | 546 | 4947517 | ||

Rab29
Genomic Map