siRNA Details
| Gene Symbol: Ipo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153774.1 | NP_722469.1 | 13 | 1475 | ||
| GenBank mRNA | AF273672.1 | - | 13 | 1420 | ||
| AF273673.1 | - | 13 | 1420 | |||
| AJ309238.1 | - | 13 | 1414 | |||
| AK036687.1 | - | 13 | 1425 | |||
| AK173127.1 | - | 13 | 1417 | |||
| BC056469.1 | - | 13 | 1442 | |||
| BC079546.1 | - | 14 | 1586 | |||
| BC098508.1 | - | 13 | 1425 | 30015513 | ||

Ipo9
Genomic Map