siRNA Details
| Gene Symbol: BC003331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077237.1 | NP_001070705.1 | 7 | 886 | ||
| NM_145511.2 | NP_663486.2 | 7 | 886 | |||
| GenBank mRNA | AK152022.1 | - | 5 | 402 | ||
| AK152955.1 | - | 5 | 402 | |||
| BC003331.1 | - | 6 | 541 | 3256634 | ||
| CB235715.1 | - | 7 | 584 | |||
| CX243768.1 | - | 3 | 134 | |||
| DQ077804.2 | - | 6 | 500 | |||

BC003331
Genomic Map