siRNA Details
| Gene Symbol: Rasal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177644.5 | NP_808312.3 | 14 | 3329 | ||
| GenBank mRNA | BC120902.2 | - | 14 | 2672 | 40086771 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasal2| Gene Symbol: Rasal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177644.5 | NP_808312.3 | 14 | 3329 | ||
| GenBank mRNA | BC120902.2 | - | 14 | 2672 | 40086771 | |