siRNA Details
| Gene Symbol: Klhl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039482.1 | NP_001034571.1 | 3 | 542 | ||
| GenBank mRNA | AK032131.1 | - | 1 | 29 | ||
| AK053102.1 | - | 3 | 492 | |||
| AK053371.1 | - | 3 | 506 | |||
| AK054169.1 | - | 3 | 510 | |||
| AK220547.1 | - | 3 | 521 | |||
| BC019571.1 | - | 3 | 514 | 4235202 | ||
| BC060183.1 | - | 3 | 471 | |||

Klhl20
Genomic Map