siRNA Details
| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146104.3 | NP_666216.1 | 4 | 653 | ||
| NM_146134.2 | NP_666246.1 | 4 | 653 | |||
| GenBank mRNA | AK050828.1 | - | 2 | 876 | ||
| AK078343.1 | - | 2 | 118 | |||
| AK148400.1 | - | 4 | 606 | |||
| AK152334.1 | - | 4 | 596 | |||
| AX775953.1 | - | 4 | 517 | |||
| BC012406.1 | - | 4 | 578 | 3483214 | ||
| BC024111.1 | - | 4 | 519 | 5098973 | ||
| BC057865.1 | - | 4 | 596 | 5320175 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014255, XM_006232977, BC087081.

Aph1a
Genomic Map