siRNA Details
| Gene Symbol: Suco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172645.2 | NP_766233.2 | 2 | 423 | ||
| GenBank mRNA | AK171856.1 | - | 2 | 362 | ||
| BC138379.1 | - | 2 | 342 | 8861399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Suco| Gene Symbol: Suco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172645.2 | NP_766233.2 | 2 | 423 | ||
| GenBank mRNA | AK171856.1 | - | 2 | 362 | ||
| BC138379.1 | - | 2 | 342 | 8861399 | ||