siRNA Details
| Gene Symbol: Rcsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038846.1 | NP_001033935.1 | 5 | 839 | ||
| NM_178593.3 | NP_848708.2 | 6 | 929 | |||
| GenBank mRNA | AK133961.1 | - | 6 | 884 | ||
| AK158429.1 | - | 6 | 870 | |||
| AK168816.1 | - | 5 | 810 | |||
| AK171323.1 | - | 819 | ||||
| AK171914.1 | - | 6 | 852 | |||
| BC025872.1 | - | 6 | 849 | 5037953 | ||

Rcsd1
Genomic Map