siRNA Details
| Gene Symbol: Fam78b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160261.1 | NP_001153733.1 | 2 | 555 | ||
| NM_001160262.1 | NP_001153734.1 | 2 | 555 | |||
| NM_175461.4 | NP_780670.2 | 2 | 555 | |||
| GenBank mRNA | AK033840.1 | - | 1 | 53 | ||
| AK047703.1 | - | 2 | 393 | |||
| AK144212.1 | - | 2 | 509 | |||
| BC066094.1 | - | 2 | 383 | |||
| BC132161.1 | - | 3 | 478 | 40130438 | ||
| BC132525.1 | - | 2 | 540 | 40130802 | ||
| CF530988.1 | - | 2 | 497 | |||

Fam78b
Genomic Map