siRNA Details
| Gene Symbol: Ndufs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153064.4 | NP_694704.1 | 11 | 1287 | ||
| GenBank mRNA | AK078474.1 | - | 11 | 1288 | ||
| AK146269.1 | - | 11 | 1292 | |||
| AK150431.1 | - | 11 | 1283 | |||
| AK151746.1 | - | 11 | 1260 | |||
| AK153364.1 | - | 11 | 1260 | |||
| AK165426.1 | - | 11 | 1283 | |||
| AK167016.1 | - | 11 | 1464 | |||
| AK168971.1 | - | 12 | 1258 | |||
| BC003898.1 | - | 11 | 1286 | |||
| BC016097.1 | - | 11 | 1274 | 4910260 | ||

Ndufs2
Genomic Map