siRNA Details
| Gene Symbol: Arhgap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005508.2 | NP_001005508.2 | 7 | 1117 | ||
| GenBank mRNA | AK031319.1 | - | 7 | 888 | ||
| AK147757.1 | - | 7 | 1124 | |||
| AK156351.1 | - | 7 | 1106 | |||
| AK220204.1 | - | 2 | 251 | |||
| BC082573.1 | - | 7 | 1065 | 30548035 | ||

Arhgap30
Genomic Map