siRNA Details
| Gene Symbol: Arhgap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005508.2 | NP_001005508.2 | 8 | 1213 | ||
| GenBank mRNA | AK031319.1 | - | 8 | 984 | ||
| AK147757.1 | - | 8 | 1220 | |||
| AK156351.1 | - | 8 | 1202 | |||
| AK220204.1 | - | 3 | 347 | |||
| BC082573.1 | - | 8 | 1161 | 30548035 | ||

Arhgap30
Genomic Map