siRNA Details
| Gene Symbol: Ifi205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172648.3 | NP_766236.2 | 3 | 437 | ||
| GenBank mRNA | AK036386.1 | - | 2 | 1364 | ||
| AK040191.1 | - | 3 | 367 | |||
| BC040425.1 | - | 3 | 437 | 3466347 | ||
| CT010211.1 | - | 2 | 285 | |||
| M74124.1 | - | 2 | 218 | |||

Ifi205
Genomic Map