siRNA Details
| Gene Symbol: Becn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290692.1 | NP_001277621.1 | 1 | 852 | ||
| GenBank mRNA | KF511582.1 | - | 1 | 811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Becn2| Gene Symbol: Becn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290692.1 | NP_001277621.1 | 1 | 852 | ||
| GenBank mRNA | KF511582.1 | - | 1 | 811 | ||