siRNA Details
Gene Symbol: Cnst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146105.3 | NP_666217.2 | 9 | 1436 | ||
GenBank mRNA | AK036332.1 | - | 9 | 1468 | ||
AK146681.1 | - | 9 | 1526 | |||
BC034107.1 | - | 2 | 454 | 4503249 | ||
BC039206.1 | - | 7 | 799 | |||
BC054802.1 | - | 9 | 1436 | 6837803 |
