siRNA Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145514.5 | NP_663489.4 | 11 | 2068 | ||
| GenBank mRNA | AK075743.1 | - | 11 | 1460 | ||
| AK082326.1 | - | 12 | 1363 | |||
| AK213047.1 | - | 2 | 60 | |||
| AK218758.1 | - | 2 | 60 | |||
| BC020044.1 | - | 7 | 813 | |||
| BC058601.1 | - | 9 | 1064 | |||
| BC094233.1 | - | 8 | 952 | |||
| BC139336.1 | - | 11 | 1230 | 8862358 | ||
| BC139337.1 | - | 11 | 1230 | 8862359 | ||

Wdr26
Genomic Map