siRNA Details
| Gene Symbol: C130074G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178692.3 | NP_848807.1 | 2 | 384 | ||
| GenBank mRNA | AK052654.1 | - | 2 | 348 | ||
| AK081755.1 | - | 2 | 363 | |||
| AK084264.1 | - | 2 | 357 | |||
| AK087640.1 | - | 2 | 375 | |||
| BC116951.1 | - | 2 | 384 | 40126270 | ||
| BC116953.1 | - | 2 | 384 | 40126272 | ||
| BY710149.1 | - | 2 | 566 | |||

C130074G19Rik
Genomic Map