siRNA Details
| Gene Symbol: Kctd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172650.2 | NP_766238.1 | 7 | 640 | ||
| GenBank mRNA | AK033491.1 | - | 7 | 641 | ||
| AK034380.1 | - | 7 | 640 | |||
| AK049362.1 | - | 7 | 638 | |||
| AK087407.1 | - | 2 | 752 | |||
| AK087865.1 | - | 7 | 641 | |||
| AK139700.1 | - | 7 | 640 | |||
| AK215667.1 | - | 2 | 53 | |||
| BC061458.1 | - | 7 | 628 | |||
| BC079581.1 | - | 7 | 627 | 5702630 | ||

Kctd3
Genomic Map