siRNA Details
| Gene Symbol: Kctd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172650.2 | NP_766238.1 | 18 | 2417 | ||
| GenBank mRNA | AK034380.1 | - | 18 | 2418 | ||
| AK049362.1 | - | 18 | 2415 | |||
| AK087865.1 | - | 18 | 2418 | |||
| AK139700.1 | - | 18 | 2421 | |||
| BC045196.1 | - | 4 | 701 | |||
| BC061458.1 | - | 18 | 2656 | |||
| BC079581.1 | - | 18 | 2404 | 5702630 | ||

Kctd3
Genomic Map