siRNA Details
| Gene Symbol: Kcnq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160139.1 | NP_001153611.1 | 3 | 924 | ||
| NM_023872.3 | NP_076361.1 | 3 | 924 | |||
| GenBank mRNA | AF263836.1 | - | 3 | 399 | ||
| AK147264.1 | - | 3 | 927 | |||
| AY679158.1 | - | 3 | 563 | |||

Kcnq5
Genomic Map