siRNA Details
| Gene Symbol: Plekhb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145516.2 | NP_663491.1 | 7, 8 | 634 | ||
| GenBank mRNA | AF189817.1 | - | 7, 8 | 594 | ||
| AK039771.1 | - | 7, 8 | 602 | |||
| AK149868.1 | - | 7, 8 | 594 | |||
| AK150074.1 | - | 7, 8 | 594 | |||
| AK166091.1 | - | 7, 8 | 602 | |||
| BC057994.1 | - | 7, 8 | 602 | 5344583 | ||

Plekhb2
Genomic Map