siRNA Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142647.1 | NP_001136119.1 | 2 | 178 | ||
| GenBank mRNA | AK030361.1 | - | 2 | 147 | ||
| AK036693.1 | - | 2 | 138 | |||
| AK048484.1 | - | 2 | 135 | |||
| AK086801.1 | - | 2 | 89 | |||
| AK170577.1 | - | 2 | 2222 | |||
| BB848894.1 | - | 2 | 181 | |||
| CD553439.1 | - | 2 | 106 | |||

Tmem194b
Genomic Map