siRNA Details
| Gene Symbol: Ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145517.4 | NP_663492.3 | 4 | 504 | ||
| GenBank mRNA | AK050385.1 | - | 4 | 489 | ||
| AK077562.1 | - | 4 | 487 | |||
| BC012315.1 | - | 4 | 459 | 3981845 | ||
| BC023695.1 | - | 4 | 484 | 5344622 | ||
| BC023903.1 | - | 4 | 447 | |||
| BC025572.1 | - | 4 | 480 | 5323819 | ||
| CN705165.1 | - | 4 | 504 | |||

Ormdl1
Genomic Map