siRNA Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160038.1 | NP_001153510.1 | 18 | 2471 | ||
| NM_001160039.1 | NP_001153511.1 | 18 | 2313 | |||
| NM_001160040.1 | NP_001153512.1 | 17 | 2367 | |||
| NM_145518.2 | NP_663493.2 | 17 | 2209 | |||
| GenBank mRNA | AK034597.1 | - | 17 | 2278 | ||
| AK036926.1 | - | 17 | 2074 | |||
| AK142711.1 | - | 17 | 2089 | |||
| AK146814.1 | - | 17 | 2227 | |||
| AK167705.1 | - | 17 | 2057 | |||
| BC006660.1 | - | 17 | 2096 | 3501043 | ||
| BC015300.1 | - | 17 | 2029 | 4236882 | ||

Ndufs1
Genomic Map