siRNA Details
| Gene Symbol: Dner | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152915.1 | NP_690879.1 | 5 | 1447 | ||
| GenBank mRNA | AB067650.1 | - | 5 | 1096 | ||
| AF370126.1 | - | 5 | 1103 | |||
| AK030642.1 | - | 5 | 1246 | |||
| AK044597.1 | - | 5 | 1210 | |||
| AY032924.1 | - | 5 | 1103 | |||
| BC034634.1 | - | 5 | 1447 | 5365190 | ||

Dner
Genomic Map