siRNA Details
| Gene Symbol: Gigyf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110212.2 | NP_001103682.1 | 11 | 1239 | ||
| NM_146112.4 | NP_666224.3 | 11 | 1257 | |||
| GenBank mRNA | AK079246.1 | - | 11 | 1209 | ||
| AK122337.1 | - | 2 | 328 | |||
| AK144058.1 | - | 11 | 1219 | |||
| AK157733.1 | - | 2 | 2331 | |||
| AK160878.1 | - | 9 | 1051 | |||
| AY176043.1 | - | 11 | 1203 | |||
| BC021370.1 | - | 11 | 1206 | |||
| BC076610.1 | - | 11 | 1202 | |||
| BC089036.1 | - | 11 | 1213 | 30695300 | ||

Gigyf2
Genomic Map