siRNA Details
| Gene Symbol: Gigyf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110212.2 | NP_001103682.1 | 10 | 985 | ||
| NM_146112.4 | NP_666224.3 | 10 | 1003 | |||
| GenBank mRNA | AK079246.1 | - | 10 | 955 | ||
| AK122337.1 | - | 1 | 74 | |||
| AK144058.1 | - | 10 | 965 | |||
| AK157733.1 | - | 1 | 2077 | |||
| AK160878.1 | - | 8 | 797 | |||
| AY176043.1 | - | 10 | 949 | |||
| BC021370.1 | - | 10 | 952 | |||
| BC076610.1 | - | 10 | 948 | |||
| BC089036.1 | - | 10 | 959 | 30695300 | ||

Gigyf2
Genomic Map