siRNA Details
| Gene Symbol: Slco4c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172658.3 | NP_766246.1 | 2, 3 | 674 | ||
| GenBank mRNA | AK049253.1 | - | 2, 3 | 674 | ||
| AK049944.1 | - | 639 | ||||
| AY654588.1 | - | 2, 3 | 674 | |||
| BC119022.1 | - | 2, 3 | 633 | 8733771 | ||

Slco4c1
Genomic Map