siRNA Details
| Gene Symbol: 2310035C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029349.1 | NP_083625.1 | 8 | 1684 | ||
| NM_173187.3 | NP_775279.2 | 9 | 1756 | |||
| GenBank mRNA | AK035724.1 | - | 8 | 1666 | ||
| AK040168.1 | - | 10 | 1392 | |||
| AK050268.1 | - | 2 | 244 | |||
| AK129368.1 | - | 8 | 1657 | |||
| AK148005.1 | - | 9 | 1736 | |||
| BC060194.1 | - | 8 | 929 | |||
| BC117949.1 | - | 9 | 1690 | 40098078 | ||
| BC117950.1 | - | 8 | 1622 | 40098082 | ||

2310035C23Rik
Genomic Map