siRNA Details
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081386.1 | NP_001074855.1 | 5 | 959 | ||
GenBank mRNA | AK148396.1 | - | 5 | 947 | ||
BC157927.1 | - | 5 | 950 | 9088100 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081386.1 | NP_001074855.1 | 5 | 959 | ||
GenBank mRNA | AK148396.1 | - | 5 | 947 | ||
BC157927.1 | - | 5 | 950 | 9088100 |