siRNA Details
| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081386.1 | NP_001074855.1 | 10 | 1707 | ||
| GenBank mRNA | BC157927.1 | - | 10 | 1698 | 9088100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh19| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081386.1 | NP_001074855.1 | 10 | 1707 | ||
| GenBank mRNA | BC157927.1 | - | 10 | 1698 | 9088100 | |