siRNA Details
| Gene Symbol: Tubb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146116.2 | NP_666228.1 | 4 | 779 | ||
| GenBank mRNA | AK002568.1 | - | 1 | 229 | ||
| AK006924.1 | - | 3 | 543 | |||
| AK076895.1 | - | 4 | 736 | |||
| AK146587.1 | - | 4 | 735 | |||
| AK167022.1 | - | 4 | 734 | |||
| AK195017.1 | - | 1 | 38 | |||
| BC005547.1 | - | 4 | 711 | 3585500 | ||
| BC022919.1 | - | 4 | 718 | 4221189 | ||
| BC083319.1 | - | 4 | 721 | 5699674 | ||
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC098458.1 | - | 2502 | |||

Tubb4b
Genomic Map