siRNA Details
Gene Symbol: Lcn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100455.2 | NP_001093925.1 | 2 | 190 | ||
GenBank mRNA | AY301267.1 | - | 2 | 190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lcn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100455.2 | NP_001093925.1 | 2 | 190 | ||
GenBank mRNA | AY301267.1 | - | 2 | 190 |