siRNA Details
| Gene Symbol: Camsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276359.1 | NP_001263288.1 | 5 | 1120 | ||
| NM_001276360.1 | NP_001263289.1 | 6 | 1153 | |||
| NM_001276361.1 | NP_001263290.1 | 6 | 1180 | |||
| GenBank mRNA | BC024715.2 | - | 5 | 835 | ||
| BC054553.1 | - | 6 | 959 | |||
| CD351317.1 | - | 5 | 601 | |||
| CF539372.1 | - | 3 | 185 | |||

Camsap1
Genomic Map